The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308097.1 | NP_001295026.1 | 6 - 7 | 754 | 154 |
NM_017769.4 | NP_060239.2 | 7 - 8 | 795 | 154 | |
XM_011536925.2 | XP_011535227.1 | 7 - 8 | 761 | 154 | |
GenBank mRNA | AB037754.1 | - | 7 - 8 | 747 | 154 |
AK093536.1 | - | 7 - 8 | 1782 | 154 | |
BC000973.2 | - | 7 - 8 | 741 | 154 |