Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 6 - 7 889 92
NM_001321121.1 NP_001308050.1 6 - 7 1000 92
NM_001321122.1 NP_001308051.1 9 - 10 1233 92
NM_002343.5 NP_002334.2 6 - 7 1000 92
GenBank mRNA AF332168.1 - 6 - 7 707 92
AK093852.1 - 9 - 10 1236 92
AK290859.1 - 6 - 7 745 92
AK292813.1 - 6 - 7 745 92
AK298035.1 - 6 - 7 889 92
AK303889.1 - 5 - 6 543 92
AK316330.1 - 6 - 7 809 92
AY137470.1 - 6 - 7 707 92
AY165046.1 - 6 - 7 719 92
AY178998.2 - 6 - 7 707 92
AY360320.1 - 6 - 7 708 92
AY493417.1 - 6 - 7 817 92
AY875691.1 - 6 - 7 707 92
BC015822.1 - 6 - 7 749 92
BC015823.1 - 6 - 7 751 92
BC022347.1 - 6 - 7 745 92
DQ522304.1 - 6 - 7 707 92
EU779935.1 - 6 - 7 707 92
EU794669.1 - 6 - 7 742 92
KF992208.1 - 6 - 7 719 92
KT006756.1 - 6 - 7 707 92
M83202.1 - 6 - 7 714 92
M83205.1 - 6 - 7 693 92
M93150.1 - 6 - 7 724 92
U07643.1 - 6 - 7 804 92
X52941.1 - 6 - 7 701 92
X53961.1 - 6 - 7 1001 92

View Full Product Details