The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005868.5 | NP_005859.1 | 2 - 3 | 311 | 146 |
GenBank mRNA | AF007551.1 | - | 2 - 3 | 264 | 146 |
AK313984.1 | - | 2 - 3 | 284 | 146 | |
BC000899.1 | - | 2 - 3 | 200 | 146 | |
BG721261.1 | - | 2 - 3 | 311 | 146 | |
BT009853.1 | - | 2 - 3 | 145 | 146 |