The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000324.2 | NP_000315.2 | 6 - 7 | 1005 | 87 |
GenBank mRNA | AB938314.1 | - | 6 - 7 | 943 | 87 |
AF031548.1 | - | 6 - 7 | 943 | 87 | |
AF178841.1 | - | 6 - 7 | 970 | 87 | |
AF179682.1 | - | 6 - 7 | 970 | 87 | |
AF179684.1 | - | 6 - 7 | 970 | 87 | |
AF179685.1 | - | 6 - 7 | 970 | 87 | |
AF187847.1 | - | 6 - 7 | 970 | 87 | |
AK313505.1 | - | 6 - 7 | 1005 | 87 | |
BC012605.1 | - | 6 - 7 | 974 | 87 | |
KJ543706.1 | - | 6 - 7 | 943 | 87 | |
X64594.1 | - | 6 - 7 | 970 | 87 |