Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHAG
Entrez Gene ID
6005
Gene Name
Rh-associated glycoprotein
Gene Aliases
CD241, OHS, OHST, RH2, RH50A, Rh50, Rh50GP, SLC42A1
Chromosome Location
Chr.6: 49605175 - 49636874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000324.2 NP_000315.2 8 - 9 1201 70
GenBank mRNA AB938314.1 - 8 - 9 1139 70
AF031548.1 - 8 - 9 1139 70
AF031549.1 - 7 - 8 1044 70
AF178841.1 - 8 - 9 1166 70
AF179682.1 - 8 - 9 1166 70
AF187847.1 - 8 - 9 1166 70
AK313505.1 - 8 - 9 1201 70
BC012605.1 - 8 - 9 1170 70
KJ543706.1 - 8 - 9 1139 70
X64594.1 - 8 - 9 1166 70

View Full Product Details