The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001872.3 | NP_001001872.2 | 1 - 2 | 162 | 77 |
NM_001320173.1 | NP_001307102.1 | 1 - 2 | 162 | 77 | |
XM_011536471.2 | XP_011534773.1 | 1 - 2 | 162 | 77 | |
XM_017021030.1 | XP_016876519.1 | 1 - 2 | 162 | 77 | |
GenBank mRNA | AK292513.1 | - | 1 - 2 | 136 | 77 |
AK292605.1 | - | 1 - 2 | 79 | 77 | |
BX161411.1 | - | 1 - 2 | 52 | 77 | |
BX161448.1 | - | 1 - 2 | 106 | 77 | |
DA126898.1 | - | 1 - 2 | 162 | 77 |