The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005729.3 | NP_005720.1 | 4 - 5 | 501 | 63 |
XM_005269379.2 | XP_005269436.1 | 4 - 5 | 499 | 63 | |
GenBank mRNA | AK312757.1 | - | 4 - 5 | 496 | 63 |
BC005020.1 | - | 4 - 5 | 487 | 63 | |
BC013204.1 | - | 4 - 5 | 478 | 63 | |
BE791220.1 | - | 4 - 5 | 517 | 63 | |
M80254.1 | - | 4 - 5 | 499 | 63 |