Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFB2M
Entrez Gene ID
64216
Gene Name
transcription factor B2, mitochondrial
Gene Aliases
Hkp1, mtTFB2
Chromosome Location
Chr.1: 246540561 - 246566263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022366.2 NP_071761.1 1 - 2 436 78
XM_011544248.2 XP_011542550.1 1 - 2 434 78
XM_017002055.1 XP_016857544.1 1 - 2 376 78
GenBank mRNA AF529366.1 - 1 - 2 311 78
AK026835.1 - 1 - 2 435 78
BC003383.1 - 1 - 2 422 78
DA038232.1 - 1 - 2 436 78

View Full Product Details