The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 16 - 17 | 2370 | 99 |
XM_006723769.2 | XP_006723832.1 | 15 - 16 | 2160 | 99 | |
GenBank mRNA | AB019219.1 | - | 16 - 17 | 2284 | 99 |
AF026031.1 | - | 16 - 17 | 2301 | 99 | |
AF221842.1 | - | 16 - 17 | 2307 | 99 | |
AK001554.1 | - | 15 - 16 | 2193 | 99 | |
AK225993.1 | - | 15 - 16 | 2204 | 99 | |
AK314310.1 | - | 16 - 17 | 2294 | 99 | |
AL137320.1 | - | 7 - 8 | 1001 | 99 | |
BC001666.2 | - | 16 - 17 | 2248 | 99 |