The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 4 - 5 | 607 | 68 |
XM_006723769.2 | XP_006723832.1 | 4 - 5 | 616 | 68 | |
GenBank mRNA | AB019219.1 | - | 4 - 5 | 521 | 68 |
AF026031.1 | - | 4 - 5 | 538 | 68 | |
AF221842.1 | - | 4 - 5 | 544 | 68 | |
AK001554.1 | - | 4 - 5 | 550 | 68 | |
AK225993.1 | - | 4 - 5 | 561 | 68 | |
AK314310.1 | - | 4 - 5 | 531 | 68 | |
BC001666.2 | - | 4 - 5 | 485 | 68 |