The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017768.4 | NP_060238.3 | 12 - 13 | 1579 | 89 |
XM_005271013.1 | XP_005271070.1 | 11 - 12 | 1406 | 89 | |
XM_011541763.1 | XP_011540065.1 | 10 - 11 | 1321 | 89 | |
XM_017001695.1 | XP_016857184.1 | 11 - 12 | 1327 | 89 | |
XM_017001696.1 | XP_016857185.1 | 9 - 10 | 1101 | 89 | |
GenBank mRNA | AK000338.1 | - | 10 - 11 | 1089 | 89 |
AK022951.1 | - | 12 - 13 | 1493 | 89 | |
AK225337.1 | - | 10 - 11 | 1089 | 89 | |
AL162076.1 | - | 2 - 3 | 154 | 89 | |
BC003407.1 | - | 12 - 13 | 1379 | 89 | |
BC008586.1 | - | 12 - 13 | 1477 | 89 |