The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 12 - 13 | 1841 | 131 |
XM_011535926.1 | XP_011534228.1 | 11 - 12 | 1709 | 131 | |
XM_011535927.2 | XP_011534229.1 | 11 - 12 | 1298 | 131 | |
XM_011535928.2 | XP_011534230.1 | 11 - 12 | 1487 | 131 | |
XM_017011003.1 | XP_016866492.1 | 11 - 12 | 1427 | 131 | |
GenBank mRNA | AJ278110.1 | - | 12 - 13 | 1704 | 131 |
AK313686.1 | - | 12 - 13 | 1541 | 131 | |
AL136751.1 | - | 12 - 13 | 1529 | 131 | |
BC066938.1 | - | 12 - 13 | 1522 | 131 |