The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 15 - 16 | 2173 | 123 |
XM_011535926.1 | XP_011534228.1 | 14 - 15 | 2041 | 123 | |
XM_011535927.2 | XP_011534229.1 | 14 - 15 | 1630 | 123 | |
XM_011535928.2 | XP_011534230.1 | 14 - 15 | 1819 | 123 | |
XM_017011003.1 | XP_016866492.1 | 14 - 15 | 1759 | 123 | |
GenBank mRNA | AJ278110.1 | - | 15 - 16 | 2036 | 123 |
AK313686.1 | - | 15 - 16 | 1873 | 123 | |
AL136751.1 | - | 15 - 16 | 1861 | 123 | |
BC066938.1 | - | 15 - 16 | 1854 | 123 | |
BQ020439.1 | - | 2 - 3 | 123 |