The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 2 - 3 | 645 | 113 |
XM_011535926.1 | XP_011534228.1 | 2 - 3 | 645 | 113 | |
XM_017011003.1 | XP_016866492.1 | 2 - 3 | 313 | 113 | |
GenBank mRNA | AJ278110.1 | - | 2 - 3 | 508 | 113 |
AK303324.1 | - | 2 - 3 | 354 | 113 | |
AL136751.1 | - | 2 - 3 | 333 | 113 | |
BC066938.1 | - | 2 - 3 | 326 | 113 | |
BX646425.1 | - | 2 - 3 | 392 | 113 |