The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 5 - 6 | 995 | 74 |
XM_011535926.1 | XP_011534228.1 | 4 - 5 | 863 | 74 | |
XM_011535927.2 | XP_011534229.1 | 4 - 5 | 452 | 74 | |
XM_011535928.2 | XP_011534230.1 | 4 - 5 | 641 | 74 | |
GenBank mRNA | AJ278110.1 | - | 5 - 6 | 858 | 74 |
AK313686.1 | - | 5 - 6 | 695 | 74 | |
AL136751.1 | - | 5 - 6 | 683 | 74 | |
BC066938.1 | - | 5 - 6 | 676 | 74 |