The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 6 - 7 | 1153 | 89 |
XM_011535926.1 | XP_011534228.1 | 5 - 6 | 1021 | 89 | |
XM_011535927.2 | XP_011534229.1 | 5 - 6 | 610 | 89 | |
XM_011535928.2 | XP_011534230.1 | 5 - 6 | 799 | 89 | |
XM_017011003.1 | XP_016866492.1 | 5 - 6 | 739 | 89 | |
GenBank mRNA | AJ278110.1 | - | 6 - 7 | 1016 | 89 |
AK303324.1 | - | 5 - 6 | 780 | 89 | |
AK313686.1 | - | 6 - 7 | 853 | 89 | |
AL136751.1 | - | 6 - 7 | 841 | 89 | |
BC066938.1 | - | 6 - 7 | 834 | 89 |