The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 7 - 8 | 1276 | 151 |
XM_011535926.1 | XP_011534228.1 | 6 - 7 | 1144 | 151 | |
XM_011535927.2 | XP_011534229.1 | 6 - 7 | 733 | 151 | |
XM_011535928.2 | XP_011534230.1 | 6 - 7 | 922 | 151 | |
XM_017011003.1 | XP_016866492.1 | 6 - 7 | 862 | 151 | |
GenBank mRNA | AJ278110.1 | - | 7 - 8 | 1139 | 151 |
AK303324.1 | - | 6 - 7 | 903 | 151 | |
AK313686.1 | - | 7 - 8 | 976 | 151 | |
AL136751.1 | - | 7 - 8 | 964 | 151 | |
BC066938.1 | - | 7 - 8 | 957 | 151 |