The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004541.3 | NP_004532.1 | 2 - 3 | 334 | 109 |
GenBank mRNA | BC000266.1 | - | 2 - 3 | 231 | 109 |
BQ680674.1 | - | 2 - 3 | 235 | 109 | |
U54993.1 | - | 2 - 3 | 226 | 109 | |
X81900.1 | - | 2 - 3 | 210 | 109 |