Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPF3B
Entrez Gene ID
65109
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
HUPF3B, MRX62, MRXS14, RENT3B, UPF3BP1, UPF3BP2, UPF3BP3, UPF3X, Upf3p-X
Chromosome Location
Chr.X: 119805311 - 119853028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023010.3 NP_075386.1 7 - 8 906 148
XM_017029739.1 XP_016885228.1 7 - 8 906 148
GenBank mRNA AK093728.1 - 7 - 8 844 148
AY013251.1 - 7 - 8 876 148
BC121017.1 - 7 - 8 849 148

View Full Product Details