Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
Chromosome Location
Chr.8: 22048955 - 22082527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114135.4 NP_001107607.1 14 - 15 1292 63
NM_001114136.2 NP_001107608.1 14 - 15 1248 63
NM_001302817.2 NP_001289746.1 12 - 13 1261 63
NM_001323378.1 NP_001310307.1 14 - 15 1381 63
NM_001323380.1 NP_001310309.1 14 - 15 1288 63
NM_001323381.1 NP_001310310.1 13 - 14 1397 63
NM_001323382.1 NP_001310311.1 14 - 15 1327 63
NM_001323388.1 NP_001310317.1 12 - 13 1128 63
NM_001323389.1 NP_001310318.1 13 - 14 1336 63
NM_001323390.1 NP_001310319.1 13 - 14 1203 63
NM_001323391.1 NP_001310320.1 13 - 14 1306 63
NM_001323392.1 NP_001310321.1 13 - 14 1467 63
NM_001323393.1 NP_001310322.1 13 - 14 1173 63
NM_001323394.1 NP_001310323.1 13 - 14 1334 63
NM_001323395.1 NP_001310324.1 13 - 14 1213 63
NM_001323396.1 NP_001310325.1 13 - 14 1217 63
NM_001323397.1 NP_001310326.1 13 - 14 1385 63
NM_001978.4 NP_001969.2 14 - 15 1542 63
XM_005273432.1 XP_005273489.1 14 - 15 1274 63
XM_005273433.1 XP_005273490.1 14 - 15 1206 63
XM_005273442.1 XP_005273499.1 13 - 14 1058 63
XM_011544435.1 XP_011542737.1 13 - 14 1467 63
XM_017013193.1 XP_016868682.1 12 - 13 1234 63
XM_017013194.1 XP_016868683.1 12 - 13 1101 63
GenBank mRNA AB209718.1 - 10 - 11 768 63
AK289650.1 - 14 - 15 1174 63
AK295452.1 - 12 - 13 1147 63
BC006318.2 - 14 - 15 1291 63
BC017445.1 - 14 - 15 1291 63
BT007396.1 - 13 - 14 1019 63
U28389.1 - 14 - 15 1469 63

View Full Product Details