Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
Chromosome Location
Chr.8: 22048955 - 22082527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114135.4 NP_001107607.1 2 - 3 288 92
NM_001114136.2 NP_001107608.1 2 - 3 244 92
NM_001114137.3 NP_001107609.1 2 - 3 418 92
NM_001114138.2 NP_001107610.1 2 - 3 405 92
NM_001302816.2 NP_001289745.1 2 - 3 538 92
NM_001323378.1 NP_001310307.1 2 - 3 377 92
NM_001323380.1 NP_001310309.1 2 - 3 284 92
NM_001323381.1 NP_001310310.1 1 - 2 393 92
NM_001323382.1 NP_001310311.1 2 - 3 323 92
NM_001323383.1 NP_001310312.1 2 - 3 244 92
NM_001323384.1 NP_001310313.1 2 - 3 377 92
NM_001323385.1 NP_001310314.1 1 - 2 393 92
NM_001323389.1 NP_001310318.1 2 - 3 377 92
NM_001323390.1 NP_001310319.1 2 - 3 244 92
NM_001978.4 NP_001969.2 2 - 3 538 92
XM_005273432.1 XP_005273489.1 2 - 3 270 92
XM_005273433.1 XP_005273490.1 2 - 3 202 92
XM_011544435.1 XP_011542737.1 2 - 3 508 92
XM_017013189.1 XP_016868678.1 2 - 3 211 92
XM_017013190.1 XP_016868679.1 2 - 3 508 92
XM_017013191.1 XP_016868680.1 2 - 3 262 92
XM_017013192.1 XP_016868681.1 2 - 3 143 92
GenBank mRNA AK055842.1 - 2 - 3 202 92
AK289650.1 - 2 - 3 170 92
BC006318.2 - 2 - 3 287 92
BC017445.1 - 2 - 3 287 92
BC052805.1 - 2 - 3 328 92
DA372459.1 - 2 - 3 270 92
DA383880.1 - 1 - 2 393 92
L19713.1 - 2 - 3 465 92
U28389.1 - 2 - 3 465 92

View Full Product Details