The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003867.3 | NP_003858.1 | 1 - 2 | 212 | 62 |
GenBank mRNA | AB009249.1 | - | 1 - 2 | 48 | 62 |
BC069475.1 | - | 1 - 2 | 48 | 62 | |
BC105131.1 | - | 1 - 2 | 86 | 62 | |
BC113489.1 | - | 1 - 2 | 86 | 62 |