The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005066.2 | NP_005057.1 | 8 - 9 | 1963 | 78 |
NR_136702.1 | 8 - 9 | 1966 | 78 | ||
NR_136703.1 | 8 - 9 | 1978 | 78 | ||
XM_005271112.4 | XP_005271169.1 | 8 - 9 | 1982 | 78 | |
XM_005271113.4 | XP_005271170.1 | 8 - 9 | 1982 | 78 | |
XM_005271115.4 | XP_005271172.1 | 8 - 9 | 1982 | 78 | |
XM_011541950.2 | XP_011540252.1 | 8 - 9 | 1982 | 78 | |
XM_017002053.1 | XP_016857542.1 | 8 - 9 | 1982 | 78 | |
XM_017002054.1 | XP_016857543.1 | 8 - 9 | 1982 | 78 | |
GenBank mRNA | BC004534.2 | - | 8 - 9 | 1651 | 78 |
BC051192.1 | - | 8 - 9 | 1965 | 78 | |
X16850.1 | - | 7 - 8 | 937 | 78 | |
X70944.1 | - | 8 - 9 | 1955 | 78 |