Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIM2
Entrez Gene ID
9447
Gene Name
absent in melanoma 2
Gene Aliases
PYHIN4
Chromosome Location
Chr.1: 159059226 - 159132351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004833.1 NP_004824.1 5 - 6 1251 142
XM_005245616.4 XP_005245673.1 4 - 5 1022 142
XM_017002848.1 XP_016858337.1 6 - 7 2478 142
GenBank mRNA AF024714.1 - 5 - 6 1251 142
AK292042.1 - 5 - 6 1160 142
BC010940.1 - 5 - 6 1253 144

View Full Product Details