Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080551.2 NP_001074020.2 17 - 18 2784 145
NM_173521.4 NP_775792.4 19 - 20 2791 145
XM_006716974.2 XP_006717037.1 21 - 22 2977 145
XM_006716975.2 XP_006717038.1 20 - 21 2872 145
XM_011518302.2 XP_011516604.1 19 - 20 4786 145
XM_011518303.1 XP_011516605.1 19 - 20 2758 145
XM_011518306.1 XP_011516608.1 16 - 17 2378 145
XM_011518309.1 XP_011516611.1 21 - 22 2977 145
XM_017014340.1 XP_016869829.1 19 - 20 3595 145
XM_017014341.1 XP_016869830.1 21 - 22 2977 145
GenBank mRNA AK126055.1 - 17 - 18 2782 145
AL833535.1 - 19 - 20 2785 145

View Full Product Details