Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080551.2 NP_001074020.2 9 - 10 1770 139
NM_173521.4 NP_775792.4 11 - 12 1777 139
NR_109816.1 13 - 14 1979 139
XM_006716974.2 XP_006717037.1 13 - 14 1963 139
XM_006716975.2 XP_006717038.1 13 - 14 1963 139
XM_011518302.2 XP_011516604.1 11 - 12 3772 139
XM_011518303.1 XP_011516605.1 11 - 12 1744 139
XM_011518306.1 XP_011516608.1 8 - 9 1364 139
XM_011518309.1 XP_011516611.1 13 - 14 1963 139
XM_017014340.1 XP_016869829.1 11 - 12 2581 139
XM_017014341.1 XP_016869830.1 13 - 14 1963 139
GenBank mRNA AK126055.1 - 9 - 10 1768 139
AL833535.1 - 11 - 12 1771 139
BC144680.1 - 13 - 14 1903 139

View Full Product Details