Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
Chromosome Location
Chr.1: 235975830 - 236065181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002508.2 NP_002499.2 2 - 3 624 110
XM_011544195.2 XP_011542497.1 2 - 3 621 110
GenBank mRNA AB209448.1 - 2 - 3 521 110
BC045606.1 - 2 - 3 534 110
M30269.1 - 2 - 3 612 110

View Full Product Details