The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000081.3 | NP_000072.2 | 9 - 10 | 4117 | 151 |
NM_001301365.1 | NP_001288294.1 | 9 - 10 | 4554 | 151 | |
XM_011544031.1 | XP_011542333.1 | 9 - 10 | 4117 | 151 | |
XM_011544032.1 | XP_011542334.1 | 9 - 10 | 4520 | 151 | |
XM_011544033.2 | XP_011542335.1 | 9 - 10 | 4267 | 151 | |
XM_011544034.1 | XP_011542336.1 | 9 - 10 | 4117 | 151 | |
XM_011544035.2 | XP_011542337.1 | 9 - 10 | 4117 | 151 | |
XM_011544036.2 | XP_011542338.1 | 5 - 6 | 1771 | 151 | |
XM_011544037.2 | XP_011542339.1 | 9 - 10 | 4117 | 151 | |
XM_011544039.2 | XP_011542341.1 | 9 - 10 | 4117 | 151 | |
XM_011544040.2 | XP_011542342.1 | 9 - 10 | 4117 | 151 | |
XM_017000150.1 | XP_016855639.1 | 9 - 10 | 4117 | 151 | |
XM_017000151.1 | XP_016855640.1 | 9 - 10 | 4117 | 151 | |
GenBank mRNA | AK307457.1 | - | 9 - 10 | 4520 | 151 |
BC144702.1 | - | 9 - 10 | 4418 | 151 | |
U67615.1 | - | 9 - 10 | 4124 | 151 | |
U70064.1 | - | 9 - 10 | 4487 | 151 | |
U72192.1 | - | 9 - 10 | 4487 | 151 | |
U84744.1 | - | 9 - 10 | 4487 | 151 |