The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167580.1 | NP_001161052.1 | 10 - 11 | 1676 | 84 |
NM_001323031.1 | NP_001309960.1 | 11 - 12 | 2209 | 84 | |
NM_001323032.1 | NP_001309961.1 | 11 - 12 | 2518 | 84 | |
NM_001323033.1 | NP_001309962.1 | 9 - 10 | 2185 | 84 | |
NM_001323036.1 | NP_001309965.1 | 10 - 11 | 1495 | 84 | |
NM_001323037.1 | NP_001309966.1 | 12 - 13 | 2425 | 84 | |
NM_001323038.1 | NP_001309967.1 | 11 - 12 | 2337 | 84 | |
NM_001323039.1 | NP_001309968.1 | 11 - 12 | 2610 | 84 | |
NM_001323040.1 | NP_001309969.1 | 11 - 12 | 1856 | 84 | |
NM_014848.5 | NP_055663.1 | 12 - 13 | 2606 | 84 | |
XM_005254998.3 | XP_005255055.1 | 11 - 12 | 2271 | 84 | |
XM_017022761.1 | XP_016878250.1 | 11 - 12 | 2222 | 84 | |
XM_017022762.1 | XP_016878251.1 | 11 - 12 | 2357 | 84 | |
GenBank mRNA | AB018278.1 | - | 10 - 11 | 1879 | 84 |
AK294902.1 | - | 10 - 11 | 1678 | 84 | |
BC030011.2 | - | 12 - 13 | 2243 | 84 | |
CR457086.1 | - | 10 - 11 | 1715 | 84 |