The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134405.1 | NP_001127877.1 | 3 - 4 | 786 | 158 |
NM_001134406.1 | NP_001127878.1 | 3 - 4 | 786 | 158 | |
NM_138290.2 | NP_612147.1 | 4 - 5 | 837 | 158 | |
XM_005250156.2 | XP_005250213.1 | 3 - 4 | 786 | 158 | |
XM_011515826.1 | XP_011514128.1 | 4 - 5 | 837 | 158 | |
XM_011515827.1 | XP_011514129.1 | 4 - 5 | 837 | 158 | |
XM_011515828.2 | XP_011514130.1 | 4 - 5 | 320 | 158 | |
XM_011515829.2 | XP_011514131.1 | 3 - 4 | 454 | 158 | |
XM_017011761.1 | XP_016867250.1 | 4 - 5 | 524 | 158 | |
XM_017011762.1 | XP_016867251.1 | 5 - 6 | 449 | 158 | |
XM_017011763.1 | XP_016867252.1 | 2 - 3 | 263 | 158 | |
XM_017011765.1 | XP_016867254.1 | 4 - 5 | 436 | 158 | |
XM_017011766.1 | XP_016867255.1 | 4 - 5 | 715 | 158 | |
GenBank mRNA | AK055233.1 | - | 4 - 5 | 835 | 158 |
AK293607.1 | - | 3 - 4 | 602 | 158 | |
AK294039.1 | - | 3 - 4 | 584 | 158 | |
AY033596.1 | - | 2 - 3 | 272 | 158 | |
BC022520.1 | - | 3 - 4 | 737 | 158 |