Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC3B
Entrez Gene ID
154661
Gene Name
RUN domain containing 3B
Gene Aliases
RPIB9, RPIP9
Chromosome Location
Chr.7: 87627656 - 87832298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134405.1 NP_001127877.1 4 - 5 873 119
NM_001134406.1 NP_001127878.1 4 - 5 873 119
NM_138290.2 NP_612147.1 5 - 6 924 119
XM_005250156.2 XP_005250213.1 4 - 5 873 119
XM_005250158.2 XP_005250215.1 2 - 3 348 119
XM_011515826.1 XP_011514128.1 5 - 6 924 119
XM_011515827.1 XP_011514129.1 5 - 6 924 119
XM_011515828.2 XP_011514130.1 5 - 6 407 119
XM_011515829.2 XP_011514131.1 4 - 5 541 119
XM_017011761.1 XP_016867250.1 5 - 6 611 119
XM_017011762.1 XP_016867251.1 6 - 7 536 119
XM_017011763.1 XP_016867252.1 3 - 4 350 119
XM_017011764.1 XP_016867253.1 2 - 3 348 119
XM_017011765.1 XP_016867254.1 5 - 6 523 119
XM_017011766.1 XP_016867255.1 5 - 6 802 119
GenBank mRNA AK055233.1 - 5 - 6 922 119
AK293607.1 - 4 - 5 689 119
AK294039.1 - 4 - 5 671 119
AY033596.1 - 3 - 4 359 119
BC022520.1 - 4 - 5 824 119

View Full Product Details