The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134405.1 | NP_001127877.1 | 5 - 6 | 958 | 79 |
NM_001134406.1 | NP_001127878.1 | 5 - 6 | 958 | 79 | |
NM_138290.2 | NP_612147.1 | 6 - 7 | 1009 | 79 | |
XM_005250156.2 | XP_005250213.1 | 5 - 6 | 958 | 79 | |
XM_005250158.2 | XP_005250215.1 | 3 - 4 | 433 | 79 | |
XM_011515826.1 | XP_011514128.1 | 6 - 7 | 1009 | 79 | |
XM_011515827.1 | XP_011514129.1 | 6 - 7 | 1009 | 79 | |
XM_011515828.2 | XP_011514130.1 | 6 - 7 | 492 | 79 | |
XM_011515829.2 | XP_011514131.1 | 5 - 6 | 626 | 79 | |
XM_017011761.1 | XP_016867250.1 | 6 - 7 | 696 | 79 | |
XM_017011762.1 | XP_016867251.1 | 7 - 8 | 621 | 79 | |
XM_017011763.1 | XP_016867252.1 | 4 - 5 | 435 | 79 | |
XM_017011764.1 | XP_016867253.1 | 3 - 4 | 433 | 79 | |
XM_017011765.1 | XP_016867254.1 | 6 - 7 | 608 | 79 | |
XM_017011766.1 | XP_016867255.1 | 6 - 7 | 887 | 79 | |
GenBank mRNA | AK055233.1 | - | 6 - 7 | 1007 | 79 |
AK293607.1 | - | 5 - 6 | 774 | 79 | |
AK294039.1 | - | 5 - 6 | 756 | 79 | |
AY033596.1 | - | 4 - 5 | 444 | 79 | |
BC022520.1 | - | 5 - 6 | 909 | 79 |