Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBD1
Entrez Gene ID
4152
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
CXXC3, PCM1, RFT
Chromosome Location
Chr.18: 50266882 - 50281774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204136.1 NP_001191065.1 12 - 13 1887 79
NM_001204137.1 NP_001191066.1 13 - 14 1962 79
NM_001204138.1 NP_001191067.1 13 - 14 1959 79
NM_001204139.1 NP_001191068.1 12 - 13 1887 79
NM_001204140.1 NP_001191069.1 12 - 13 1794 79
NM_001204141.1 NP_001191070.1 11 - 12 1737 79
NM_001323942.1 NP_001310871.1 13 - 14 1962 79
NM_001323949.1 NP_001310878.1 13 - 14 1954 79
NM_001323952.1 NP_001310881.1 11 - 12 1644 79
NM_001323954.1 NP_001310883.1 10 - 11 1650 79
NM_015844.2 NP_056669.2 11 - 12 1719 79
NM_015846.3 NP_056671.2 12 - 13 1887 79
NM_015847.3 NP_056723.2 11 - 12 1740 79
XM_005258271.2 XP_005258328.1 12 - 13 1634 79
XM_006722456.2 XP_006722519.1 13 - 14 1709 79
XM_011525993.2 XP_011524295.1 13 - 14 1709 79
XM_011525994.2 XP_011524296.1 13 - 14 1709 79
XM_011525998.2 XP_011524300.1 12 - 13 1631 79
XM_011526001.1 XP_011524303.1 11 - 12 1466 79
XM_017025753.1 XP_016881242.1 12 - 13 1634 79
XM_017025755.1 XP_016881244.1 12 - 13 1634 79
XM_017025760.1 XP_016881249.1 12 - 13 1634 79
XM_017025765.1 XP_016881254.1 11 - 12 1466 79
XM_017025766.1 XP_016881255.1 12 - 13 1541 79
XM_017025767.1 XP_016881256.1 12 - 13 1541 79
XM_017025768.1 XP_016881257.1 11 - 12 1466 79
XM_017025770.1 XP_016881259.1 11 - 12 1466 79
XM_017025771.1 XP_016881260.1 11 - 12 1466 79
XM_017025773.1 XP_016881262.1 10 - 11 1397 79
XM_017025774.1 XP_016881263.1 10 - 11 1397 79
GenBank mRNA AF078830.1 - 11 - 12 1450 79
AF078832.1 - 11 - 12 1282 79
AK225889.1 - 12 - 13 1797 79
AK291519.1 - 12 - 13 1888 79
AK295401.1 - 13 - 14 1694 79
AK302004.1 - 13 - 14 1654 79
BC033242.1 - 11 - 12 1445 79
BC047353.1 - 11 - 12 1422 79

View Full Product Details