Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANKRD26P1
- Entrez Gene ID
-
124149
- Gene Name
- ankyrin repeat domain 26 pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 46469337 - 46569097 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026556.1 |
|
2 - 3 |
553 |
123 |
GenBank mRNA |
BC070117.1 |
- |
2 - 3 |
553 |
123 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
EL736909.1 |
- |
2 - 3 |
371 |
|
Chr. 16 - 46559954 - 46568915 [-] on Build GRCh38 |
123 |
GenBankmRNA |
BF377398.1 |
- |
2 - 3 |
154 |
|
Chr. 16 - 46563865 - 46568697 [-] on Build GRCh38 |
123 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC400533 |
FLJ26245 |
|
-10.8mb
|
Anti-sense |
Chr. - 35746552 - 35756624 on Build GRCh38 |
Downstream |
SHC binding and spindle associated 1 |
SHCBP1 |
PAL |
16.3kb
|
Sense |
Chr. - 46578591 - 46624933 on Build GRCh38 |
View Full Product Details