Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSTD2
Entrez Gene ID
158427
Gene Name
thiosulfate sulfurtransferase like domain containing 2
Gene Aliases
C9orf97
Chromosome Location
Chr.9: 97600080 - 97633680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139246.4 NP_640339.4 5 - 6 1216 80
XM_011518312.2 XP_011516614.1 4 - 5 679 80
XM_017014342.1 XP_016869831.1 4 - 5 741 80
GenBank mRNA AK056453.1 - 3 - 4 343 80
AK094043.1 - 5 - 6 929 80
AK122809.1 - 3 - 4 1330 80
AK128615.1 - 6 - 7 882 80
AK291199.1 - 5 - 6 837 80
BC035604.1 - 5 - 6 863 80
BX538308.1 - 5 - 6 857 80
BX648493.1 - 5 - 6 855 80
KU178877.1 - 4 - 5 647 80
KU178878.1 - 4 - 5 619 80

View Full Product Details