Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
Chromosome Location
Chr.7: 151175698 - 151187832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142460.1 NP_001135932.2 3 - 4 1362 66
GenBank mRNA AF417920.1 - 3 - 4 1364 66

View Full Product Details