Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
Chromosome Location
Chr.7: 151175698 - 151187832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142459.1 NP_001135931.2 5 - 6 1659 71
NM_001142460.1 NP_001135932.2 4 - 5 1545 71
NM_080871.3 NP_543147.2 5 - 6 1463 71
GenBank mRNA AF417920.1 - 4 - 5 1547 71
AK055536.1 - 5 - 6 1369 71
AK125565.1 - 5 - 6 1463 71
BC126351.1 - 5 - 6 1423 71

View Full Product Details