Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
Chromosome Location
Chr.2: 231198546 - 231394991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271466.2 NP_001258395.1 8 - 9 918 62
NM_001291656.1 NP_001278585.1 9 - 10 1071 62
NM_025139.5 NP_079415.3 9 - 10 1073 62
XM_011511905.1 XP_011510207.1 9 - 10 991 62
XM_011511906.1 XP_011510208.1 9 - 10 991 62
XM_011511908.2 XP_011510210.1 9 - 10 991 62
XM_011511909.2 XP_011510211.1 9 - 10 991 62
XM_011511910.1 XP_011510212.1 9 - 10 991 62
XM_011511911.1 XP_011510213.1 9 - 10 991 62
XM_011511912.1 XP_011510214.1 9 - 10 991 62
XM_011511913.2 XP_011510215.1 9 - 10 991 62
XM_011511914.2 XP_011510216.1 9 - 10 991 62
XM_011511915.2 XP_011510217.1 9 - 10 991 62
XM_011511916.2 XP_011510218.1 9 - 10 991 62
XM_017005018.1 XP_016860507.1 9 - 10 1249 62
XM_017005019.1 XP_016860508.1 9 - 10 1167 62
XM_017005020.1 XP_016860509.1 9 - 10 1249 62
XM_017005021.1 XP_016860510.1 9 - 10 991 62
XM_017005022.1 XP_016860511.1 9 - 10 991 62
XM_017005023.1 XP_016860512.1 9 - 10 1249 62
XM_017005025.1 XP_016860514.1 9 - 10 991 62
XM_017005026.1 XP_016860515.1 9 - 10 1190 62
GenBank mRNA AK022646.1 - 7 - 8 775 62
AY219922.1 - 9 - 10 952 62
AY929062.1 - 9 - 10 945 62
BC004514.2 - 5 - 6 495 62
BC065271.1 - 8 - 9 905 62
BX537956.1 - 9 - 10 989 62

View Full Product Details