The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204872.1 | NP_001191801.1 | 7 - 8 | 864 | 59 |
NM_001204873.1 | NP_001191802.1 | 7 - 8 | 814 | 59 | |
NM_024663.3 | NP_078939.3 | 6 - 7 | 921 | 59 | |
GenBank mRNA | AB075854.1 | - | 7 - 8 | 2211 | 59 |
AK021645.1 | - | 6 - 7 | 861 | 59 | |
AK223094.1 | - | 6 - 7 | 879 | 59 | |
AK297667.1 | - | 7 - 8 | 810 | 59 | |
AK307771.1 | - | 7 - 8 | 859 | 59 | |
AK315684.1 | - | 6 - 7 | 853 | 59 | |
AL833971.1 | - | 5 - 6 | 765 | 59 | |
BC020507.1 | - | 6 - 7 | 887 | 59 |