The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006295.2 | NP_006286.1 | 3 - 4 | 948 | 72 |
XM_005249362.2 | XP_005249419.1 | 3 - 4 | 941 | 72 | |
GenBank mRNA | BC012808.2 | - | 3 - 4 | 718 | 72 |
CX167000.1 | - | 2 - 3 | 464 | 72 | |
X59303.1 | - | 3 - 4 | 744 | 72 |