The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144889.1 | NP_001138361.1 | 11 - 12 | 1652 | 64 |
NM_001144890.1 | NP_001138362.1 | 11 - 12 | 1676 | 64 | |
NM_144712.4 | NP_653313.3 | 10 - 11 | 1301 | 64 | |
GenBank mRNA | AK298256.1 | - | 11 - 12 | 1562 | 64 |
AK298277.1 | - | 11 - 12 | 1585 | 64 |