Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNRD1
Entrez Gene ID
7296
Gene Name
thioredoxin reductase 1
Gene Aliases
GRIM-12, TR, TR1, TRXR1, TXNR
Chromosome Location
Chr.12: 104215779 - 104350307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093771.2 NP_001087240.1 13 - 14 1567 117
NM_001261445.1 NP_001248374.1 11 - 12 1989 117
NM_001261446.1 NP_001248375.1 10 - 11 1544 117
NM_003330.3 NP_003321.3 11 - 12 1904 117
NM_182729.2 NP_877393.1 11 - 12 1667 117
NM_182742.2 NP_877419.1 10 - 11 1794 117
NM_182743.2 NP_877420.1 10 - 11 1557 117
GenBank mRNA AF077367.1 - 10 - 11 1221 117
AF208018.1 - 9 - 10 1117 117
AJ001050.1 - 9 - 10 1130 117
AK128515.1 - 10 - 11 1452 117
AK129977.1 - 2 - 3 237 117
AK226071.1 - 11 - 12 1375 117
AK293322.1 - 10 - 11 1204 117
AK295067.1 - 11 - 12 1303 117
AK296495.1 - 10 - 11 1194 117
AK304241.1 - 13 - 14 1550 117
AK308881.1 - 11 - 12 1598 117
AK312262.1 - 11 - 12 1566 117
AY057105.1 - 13 - 14 1543 117
BC018122.1 - 11 - 12 1280 117
BT019640.1 - 9 - 10 1093 117
CR536506.1 - 9 - 10 1093 117
D88687.1 - 11 - 12 1345 117
S79851.1 - 11 - 12 1532 117
X91247.1 - 11 - 12 1532 117

View Full Product Details