The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324512.1 | NP_001311441.1 | 17 - 18 | 4055 | 81 |
NM_030955.3 | NP_112217.2 | 19 - 20 | 4310 | 81 | |
XM_017009905.1 | XP_016865394.1 | 20 - 21 | 4409 | 81 | |
XM_017009906.1 | XP_016865395.1 | 19 - 20 | 3810 | 81 | |
XM_017009907.1 | XP_016865396.1 | 12 - 13 | 2702 | 81 | |
XM_017009908.1 | XP_016865397.1 | 9 - 10 | 2294 | 81 | |
XM_017009909.1 | XP_016865398.1 | 8 - 9 | 2208 | 81 | |
GenBank mRNA | BC131733.1 | - | 17 - 18 | 3804 | 81 |