The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005271112.4 | XP_005271169.1 | 10 - 11 | 5553 | 85 |
XM_005271113.4 | XP_005271170.1 | 10 - 11 | 5553 | 85 |