Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
Chromosome Location
Chr.2: 27364352 - 27370457 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034116.1 NP_001029288.1 6 - 7 736 70
NM_001318965.1 NP_001305894.1 5 - 6 695 70
NM_001318966.1 NP_001305895.1 6 - 7 786 70
NM_001318967.1 NP_001305896.1 6 - 7 729 70
NM_001318968.1 NP_001305897.1 5 - 6 625 70
NM_015636.3 NP_056451.3 6 - 7 733 70
NM_172195.3 NP_751945.2 5 - 6 692 70
XM_006712132.1 XP_006712195.1 6 - 7 783 70
GenBank mRNA AF057699.1 - 6 - 7 603 70
AF112207.1 - 6 - 7 612 70
AF218009.1 - 6 - 7 757 70
AK300734.1 - 6 - 7 783 70
AL050109.1 - 3 - 4 206 70
BC001870.1 - 6 - 7 594 70
BC091502.1 - 6 - 7 609 70

View Full Product Details