Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPV17
Entrez Gene ID
4358
Gene Name
MPV17, mitochondrial inner membrane protein
Gene Aliases
MTDPS6, SYM1
Chromosome Location
Chr.2: 27309492 - 27323102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BG714998.1 - 4 - 5 281 85

View Full Product Details