Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN13
Entrez Gene ID
27075
Gene Name
tetraspanin 13
Gene Aliases
NET-6, NET6, TM4SF13
Chromosome Location
Chr.7: 16753726 - 16784536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014399.3 NP_055214.1 1 - 2 301 80
GenBank mRNA AF100759.1 - 1 - 2 221 80
AF120265.1 - 1 - 2 127 80
AK026587.1 - 1 - 2 265 80
AK075475.1 - 1 - 2 245 80
AK316595.1 - 1 - 2 244 80
AY358995.1 - 1 - 2 232 80
BC033863.1 - 1 - 2 153 80
CR542107.1 - 1 - 2 59 80

View Full Product Details