Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDRGK1
Entrez Gene ID
65992
Gene Name
DDRGK domain containing 1
Gene Aliases
C20orf116, UFBP1, dJ1187M17.3
Chromosome Location
Chr.20: 3190350 - 3204697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023935.2 NP_076424.1 6 - 7 747 108
GenBank mRNA BC000643.1 - 6 - 7 699 108
BC007957.1 - 6 - 7 705 108
BC011851.1 - 6 - 7 700 108
CB305409.1 - 3 - 4 108

View Full Product Details