The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152740.3 | NP_689953.1 | 3 - 4 | 574 | 105 |
XM_017011719.1 | XP_016867208.1 | 4 - 5 | 604 | 105 | |
GenBank mRNA | AB050000.1 | - | 3 - 4 | 448 | 105 |
AF529362.1 | - | 3 - 4 | 361 | 105 | |
AK025558.1 | - | 2 - 3 | 186 | 105 | |
AK316605.1 | - | 3 - 4 | 379 | 105 | |
BC020167.1 | - | 3 - 4 | 435 | 105 | |
BC032324.1 | - | 3 - 4 | 459 | 105 |