The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012213.2 | NP_036345.2 | 4 - 5 | 964 | 85 |
GenBank mRNA | AF090834.1 | - | 4 - 5 | 944 | 85 |
AF153679.1 | - | 4 - 5 | 922 | 85 | |
BC000286.1 | - | 4 - 5 | 916 | 85 | |
BC052592.1 | - | 4 - 5 | 964 | 85 |